Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM96B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576272
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Signs of Rubinstein-Taybi include mental retardation and predisposition to tumor growth and white blood cell neoplasias. Crohn's disease is a gastrointestinal inflammatory condition associated with chromosome 16 through the NOD2 gene. An association with systemic lupus erythematosis and a number of other autoimmune disorders with the pericentromeric region of chromosome 16 has led to the identification of SLC5A11 as a potential autoimmune modifier. The FAM96B gene product has been provisionally designated FAM96B pending further characterization.
Specifications
FAM96B | |
Polyclonal | |
Unconjugated | |
CIAO2B | |
1110019N10Rik; CGI-128; CIAB; Ciao2b; Cytosolic iron-sulfur assembly component 2B; Fam96b; family with sequence similarity 96 member B; family with sequence similarity 96, member B; HSPC118; MIP18; Mitotic spindle-associated MMXD complex subunit MIP18; MSS19-interacting protein of 18 kDa; Protein FAM96B; RGD1311762 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51647, 68523 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9D187, Q9Y3D0 | |
CIAO2B | |
Recombinant fusion protein of human FAM96B. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction