Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM98A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | FAM98A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1567820
![]() |
Novus Biologicals
NBP15678120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156781
![]() |
Novus Biologicals
NBP156781 |
100 μL |
Each for $487.50
|
|
|||||
Description
FAM98A Polyclonal specifically detects FAM98A in Human samples. It is validated for Western Blot.Specifications
FAM98A | |
Polyclonal | |
Rabbit | |
Q8NCA5 | |
25940 | |
Synthetic peptides corresponding to FAM98A(family with sequence similarity 98, member A) The peptide sequence was selected from the middle region of FAM98A. Peptide sequence QKRQDGPQQQTGGRGGGRGGYEHSSYGGRGGHEQGGGRGGRGGYDHGGRG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DKFZP564F0522, DKFZp686O03192, family with sequence similarity 98, member A, hypothetical protein LOC25940 | |
FAM98A | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title