Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM98A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590721 Supplier Invitrogen™ Supplier No. PA590721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MECDLMETDI LESLEDLGYK GPLLEDGALS QAVSAGASSP EFTKLCAWLV SELRVLCKLE ENVQATNSPS EAEEFQLEVS GLLGEMNCPY LSLTSGDVTK RLLIQKNCLL LLTYLISELE AARMLCVNAP PKKAQEGGGS EVFQELKGIC IALGMSKPPA NITMFQFFSG IEKKLKETLA KVPPNHVGKP LLKKPMGPAH WEKIEAINQA IANEYEVRRK; Positive Samples: U-87MG, LO2, A-549.

FAM98A belongs to the FAM98 family. The function of FAM98A remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM98A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Fam98a
Gene Accession No. Q3TJZ6, Q5FWT1, Q8NCA5
Gene Alias 2810405J04Rik; AL024114; Fam98a; FAM98A-like protein; family with sequence similarity 98 member A; family with sequence similarity 98, member A; protein FAM98A; RGD1305486
Gene Symbols Fam98a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human FAM98A (NP_0562903).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25940, 313873, 72722
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.