Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM98A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544301
Description
Peptide sequence: YTGSGYQGGG YQQDNRYQDG GHHGDRGGGR GGRGGRGGRG GRAGQGGGWG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
FAM98A belongs to the FAM98 family. The function of FAM98A remains unknown.
Specifications
FAM98A | |
Polyclonal | |
Unconjugated | |
Fam98a | |
2810405J04Rik; AL024114; Fam98a; FAM98A-like protein; family with sequence similarity 98 member A; family with sequence similarity 98, member A; protein FAM98A; RGD1305486 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25940 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NCA5 | |
Fam98a | |
Synthetic peptide directed towards the middle region of human FAM98A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction