Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM98B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | FAM98B |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FAM98B Polyclonal specifically detects FAM98B in Human samples. It is validated for Western Blot.Specifications
| FAM98B | |
| Polyclonal | |
| Rabbit | |
| A8MUW5 | |
| 283742 | |
| Synthetic peptides corresponding to FAM98B(family with sequence similarity 98, member B) The peptide sequence was selected from the N terminal of FAM98B. Peptide sequence LTKAAEGGLSSPEFSELCIWLGSQIKSLCNLEESITSAGRDDLESFQLEI. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| family with sequence similarity 98, member B, FLJ38426 | |
| FAM98B | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title