Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM98B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552544 Supplier Invitrogen™ Supplier No. PA552544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STTSDIPHML NQVESKVKDI LSKVQKNHVG KPLLKMDLNS EQAEQLERIN DALSCEYECR RRMLMKRLDV TVQSFGWSDR AKVKTDDIAR IYQPKRYALS PKTTITMAHL LAAREDLSKI IRTSSGTSRE KTACAI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 69%.

FAM98B positively stimulates PRMT1-induced protein arginine dimethylated arginine methylation. Promotes colorectal cancer cell malignancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM98B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM98B
Gene Accession No. Q52LJ0
Gene Alias 2610510H03Rik; Fam98b; family with sequence similarity 98 member B; family with sequence similarity 98, member B; protein FAM98B; RGD1564603
Gene Symbols FAM98B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM98B. Recombinant protein control fragment (Product #RP-89734).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283742
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.