Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM98B Polyclonal Antibody

FAM98B Polyclonal Antibody
Supplier: Thermo Scientific PA552568
Description
FAM98B Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| FAM98B | |
| Polyclonal | |
| Unconjugated | |
| FAM98B | |
| 2610510H03Rik; Fam98b; family with sequence similarity 98 member B; family with sequence similarity 98, member B; protein FAM98B; RGD1564603 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 283742, 499866, 68215 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q52LJ0, Q80VD1 | |
| FAM98B | |
| Recombinant protein corresponding to Human FAM98B. Recombinant protein control fragment (Product #RP-89733). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction