Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM98C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559356 Supplier Invitrogen™ Supplier No. PA559356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPGAGLRLLR FLCSELQATR LLCLRSLLDP SPRPPLGEGV VEGAGMVQEL DLTLQALGLP RPAPGTPASQ LLQELHA Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.

FAM98C has two isoforms with MW 28-30 kDa and 37-40 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM98C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM98C
Gene Accession No. Q17RN3
Gene Alias 1110006G06Rik; AI836081; AV026483; B230110F21Rik; FAM98C; family with sequence similarity 98 member C; family with sequence similarity 98, member C; protein FAM98C; RGD1303117
Gene Symbols FAM98C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM98C. Recombinant protein control fragment (Product #RP-99064).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147965
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.