Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557171 Supplier Invitrogen™ Supplier No. PA557171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEEEGEEEEL IRIFQEQQKK WQQYRSVRRE RLKEMKLLRD QFVKALEDFE DLCDRVFSDE DSEL Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 45%.

The FAM9B gene, also known as Family With Sequence Similarity 9 Member B, is located on the X chromosome at Xp22.3. This gene is specifically expressed in the testes and is involved in meiotic processes crucial for gametogenesis. FAM9B encodes a protein that localizes to meiotic chromosome cores, playing a role in the regulation and stabilization of chromosome segregation during meiosis. Its expression pattern suggests a fundamental role in the generation of gametes, and defects in this gene could have implications for male fertility. The detailed mechanisms by which FAM9B functions are still under investigation, but its association with meiosis highlights its importance in reproductive biology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM9B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM9B
Gene Accession No. Q8IZU0
Gene Alias FAM9B; family with sequence similarity 9 member B; family with sequence similarity 9, member B; protein FAM9B; testis expressed 39B; TEX39B
Gene Symbols FAM9B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM9B. Recombinant protein control fragment (Product #RP-95467).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171483
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.