Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Famotidine
SDP

Catalog No. 7290100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-901-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 72-901-00 Supplier R&D Systems™ Supplier No. 7290/100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and high affinity H2 biased inverse agonist

Famotidine is a potent and high affinity histamine H2 receptor biased inverse agonist (Kd = 14 nM, IC50 = 33 nM). Reduces basal receptor activity by 75%. Inhibits Gs-mediated pathways but acts as a partial agonist of β-arrestin recruitment (EC50 = 105 nM). Inhibits gastric acid secretion and histamine-induced mast cell activation. Also inhibits HIV replication in vitro.

Specifications

Chemical Name or Material 3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-N-(aminosulfonyl)propanimidamide
CAS 76824-35-6
Quantity 100 mg
Product Line Tocris
Target Histamine H2 Receptor Antagonists
Molecular Formula C8H15N7O2S3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.