Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521512 Supplier Invitrogen™ Supplier No. PA521512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: FAN tansfected 293T cell. Predicted reactivity: Mouse (93%), Rat (91%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a WD-repeat protein that binds the cytoplasmic sphingomyelinase activation domain of the 55kD tumor necrosis factor receptor. This protein is required for TNF-mediated activation of neutral sphingomyelinase and may play a role in regulating TNF-induced cellular responses such as inflammation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene NSMAF
Gene Accession No. Q92636
Gene Alias AA959567; C630007J05; factor associated with neutral sphingomyelinase activation; Factor associated with N-SMase activation; FAN; GRAMD5; neutral sphingomyelinase (N-SMase) activation associated factor; neutral sphingomyelinase activation associated factor; NSMAF; Protein FAN
Gene Symbols NSMAF
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FAN. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.