Learn More
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2).
- Fanconi anemia, complementation group A
- Transfected cell line: 293T
- Plasmid: pCMV-FANCA full-length
- Host: Human
- Theoretical molecular weight: 32.78kDa
- Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | FANCA 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC008979 |
| Molecular Weight (g/mol) | 32.78 |
| pH Range | 6.8 |
| Plasmid | pCMV-FANCA full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-FANCA antibody (H00002175-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.