Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCB Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119087UV
Description
FANCB Polyclonal antibody specifically detects FANCB in Human samples. It is validated for Western BlotSpecifications
FANCB | |
Polyclonal | |
Western Blot | |
EC 2.8.1, EC 3.6.3.14, FA2, FAAP90, FAAP95FAB, FACB, Fanconi anemia group B protein, Fanconi anemia, complementation group B, Fanconi anemia-associated polypeptide of 95 kDa, FLJ34064, Protein FACB | |
The immunogen recognized by this antibody maps to a region between residue 820 and 859 of human Fanconi anemia complementation group B using the numbering given in entry NP_001018123.1 (GeneID 2187). | |
0.1 mL | |
Cancer, DNA Repair, Epigenetics | |
2187 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction