Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCB Antibody (M38P3E10), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252406AF594
Description
FANCB Monoclonal specifically detects FANCB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FANCB | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
FANCB | |
Ovalbumin-conjugated synthetic peptide- DRRENIHPYR | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
M38P3E10 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.8.1, EC 3.6.3.14, FA2, FAAP90, FAAP95FAB, FACB, Fanconi anemia group B protein, Fanconi anemia, complementation group B, Fanconi anemia-associated polypeptide of 95 kDa, FLJ34064, Protein FACB | |
Mouse | |
Protein A purified | |
Cancer, DNA Repair, Epigenetics | |
2187 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction