Learn More
FANCB Antibody (M38P3E10), Alexa Fluor™ 750, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP252406AF750
Catalog No. NP22406A70
Only null left
Add to Cart
Description
Description
FANCB Monoclonal specifically detects FANCB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
FANCB
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
FANCB
Ovalbumin-conjugated synthetic peptide- DRRENIHPYR
0.1 mL
Primary
Human
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
M38P3E10
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
EC 2.8.1, EC 3.6.3.14, FA2, FAAP90, FAAP95FAB, FACB, Fanconi anemia group B protein, Fanconi anemia, complementation group B, Fanconi anemia-associated polypeptide of 95 kDa, FLJ34064, Protein FACB
Mouse
Protein A purified
Cancer, DNA Repair, Epigenetics
2187
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.