Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FANCB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551829
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551829 Supplier Invitrogen™ Supplier No. PA551829
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSLNSDCLTS FKITDLGKIN YSSEPSDCNE DDLFEDKQEN RYLVVPPLET GLKVCFSSFR ELRQHLLLKE KIISKSYKAL INLVQGKDDN TSSAEEKECL VPLCGEEENS VHILDEKLSD NFQDSEQLVE KIWY Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 46%.

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group B. Alternative splicing results in two transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FANCB
Gene Accession No. Q8NB91
Gene Alias FA2; FAAP90; FAAP95; FAB; FACB; FANCB; Fanconi anemia complementation group B; Fanconi anemia group B protein; Fanconi anemia-associated polypeptide of 95 kDa; FLJ34064; Protein FACB
Gene Symbols FANCB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FANCB. Recombinant protein control fragment (Product #RP-92249).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.