Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB1002565
Description
FANCC Polyclonal specifically detects FANCC in Human samples. It is validated for Western Blot.Specifications
FANCC | |
Polyclonal | |
Western Blot 1:1000 | |
Q00597 | |
FANCC | |
A C-terminal peptide within residues 450-558 of human FANCC. [SwissProt# Q00597] | |
Unpurified | |
RUO | |
Primary | |
Human. Immunogen sequence has 92% homology to rat and mouse and 81% homology to bovine. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
Whole antisera with No Preservative | |
FA3, FACCProtein FACC, FACFanconi anemia group C protein, Fanconi anemia, complementation group C, FLJ14675 | |
Rabbit | |
65 kDa | |
0.1 mL | |
Breast Cancer, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
2176 | |
Human | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction