Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCC Antibody, Novus Biologicals™
SDP

Catalog No. NB1002565SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002565SS 0.025 mL
NB1002565 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002565SS Supplier Novus Biologicals Supplier No. NB1002565SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

FANCC Polyclonal specifically detects FANCC in Human samples. It is validated for Western Blot.

Specifications

Antigen FANCC
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation Whole antisera with No Preservative
Gene Accession No. Q00597
Gene Alias FA3, FACCProtein FACC, FACFanconi anemia group C protein, Fanconi anemia, complementation group C, FLJ14675
Gene Symbols FANCC
Host Species Rabbit
Immunogen A C-terminal peptide within residues 450-558 of human FANCC. [SwissProt# Q00597]
Molecular Weight of Antigen 65 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 2176
Test Specificity Human. Immunogen sequence has 92% homology to rat and mouse and 81% homology to bovine.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.