Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRP78/HSPA5 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $499.50

Specifications

Antigen GRP78/HSPA5
Concentration 1.0 mg/ml
Dilution Western Blot 0.5 ug/ml, Simple Western 1:25, Immunocytochemistry/Immunofluorescence 1:50
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP106277SS
SDP
View Documents
Novus Biologicals
NBP106277SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP106277
SDP
View Documents
Novus Biologicals
NBP106277
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

GRP78/HSPA5 Polyclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

GRP78/HSPA5
Western Blot 0.5 ug/ml, Simple Western 1:25, Immunocytochemistry/Immunofluorescence 1:50
Polyclonal
Rabbit
Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response
PBS and 30% Glycerol with 0.1% Sodium Azide
3309
Synthetic peptide made to a C-terminal portion of rat GRP78 (within residues 600-654). [Swiss-Prot# P06761]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
78 kDa
1.0 mg/ml
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse, Rat
78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2
HSPA5
IgG
Affinity Purified
Mouse and rat. Reacts weakly with human.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.