Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2 Antibody (1290C) - BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$241.00 - $482.50

Specifications

Antigen FANCD2
Clone 1290C
Concentration 1.0mg/mL
Dilution Western Blot 0.5μg/ml, Knockout Validated
Applications KnockDown, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP261930SS
SDP
View Documents
Novus Biologicals
NBP261930SS
0.025 mg
Each for $241.00
Add to Cart
 
NBP261930
SDP
View Documents
Novus Biologicals
NBP261930
0.1 mg
Each for $482.50
Add to Cart
 
Description

Description

FANCD2 Monoclonal specifically detects FANCD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.
Specifications

Specifications

FANCD2
1.0mg/mL
KnockDown, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
FANCD2
IgG
Protein A or G purified
164.1 kDa
1290C
Western Blot 0.5μg/ml, Knockout Validated
Monoclonal
Purified
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
PBS with 0.02% Sodium Azide
2177
This FANCD2 Antibody (1290C) was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit