Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCD2 Antibody (1290C), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP261930JF549
This item is not returnable.
View return policy
Description
FANCD2 Monoclonal antibody specifically detects FANCD2 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDownSpecifications
FANCD2 | |
Monoclonal | |
Janelia Fluor 549 | |
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
This FANCD2 Antibody (1290C) was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] | |
0.1 mL | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
2177 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, KnockDown | |
1290C | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction