Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCD2 Antibody (1290D), PE/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP254808PECY7
Description
FANCD2 Monoclonal specifically detects FANCD2 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular), Knockout Validated.Specifications
FANCD2 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
FANCD2 | |
This FANCD2 Antibody (1290D) [PE/Cy7] was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] | |
0.1 mL | |
Primary | |
Human | |
Supernatant |
Flow Cytometry, Immunoassay | |
1290D | |
Flow Cytometry, Flow (Intracellular), Knockout Validated | |
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
2177.0 | |
Store at 4°C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction