Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2 Mouse anti-Human, Unconjugated, Clone: 10F8.1, MilliporeSigma™

Catalog No. MABS1889100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1889-100 100 μg
MABS1889-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS1889-100 Supplier MilliporeSigma Supplier No. MABS1889100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FANCD2, clone 10F8.1, Cat. No. MABS1889, is a mouse monoclonal antibody that detects Fanconi anemia group D2 protein and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCD2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10F8.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias FFanconi anemia group D2 protein
Gene Symbols FANCD2;FACD
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to first 150 amino acids of human Fanconi anemia group D2 protein.
Molecular Weight of Antigen ∽166 kDa observed; 164.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_149075
Test Specificity Clone 10F8.1 specifically detects human Fanconi anemia group D2 protein. It targets an epitope with in the first 150 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.