Learn More
Invitrogen™ FANCD2 Polyclonal Antibody, DyLight™ 550
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA522708
Catalog No. PIPA522708
Only null left
Add to Cart
Description
Description
In Western blot, this antibody should recognize a band at ~166 kDa (post-translationally modified form). Additional bands may be seen at lower molecular weights. In immunofluorescence, this has been tested in human MMC and IR-treated MEF cells.
FANCD2 (Fanconi anemia group D2 protein) is required for maintenance of chromosomal stability. It promotes accurate and efficient pairing of homologs during meiosis. FANCD2 is involved in the repair of DNA double-stranded breaks, both by homologous recombination and single-stranded annealing. It is required for the targeting/stabilization of BLM to non-centromeric abnormal structures induced by replicative stress. FANCD2 promotes BRCA2/FANCD1 loading onto damaged chromatin. Defects in the FANCD2 gene result in Fanconi anemia complementation group D2. This disorder affects all bone marrow elements resulting in anemia, leukopenia, and thrombopenia.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FANCD2
Polyclonal
DyLight 550
FANCD2
2410150O07Rik; AU015151; BB137857; DKFZp762A223; FA4; FACD; FACD2; FAD; FAD2; FA-D2; FANCD; FANCD2; Fanconi anemia complementation group D2; Fanconi anemia D2 protein; Fanconi anemia group D2 protein; Fanconi anemia group D2 protein homolog; Fanconi anemia, complementation group D2; FLJ23826; Protein FACD2
Rabbit
Antigen affinity chromatography
RUO
211651, 2177, 312641, 394241, 484659
4°C, store in dark
Liquid
ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
0.8 mg/mL
50mM sodium borate with 0.05% sodium azide
Q6IV68, Q80V62, Q9BXW9
FANCD2
Human FANCD2 fusion protein (N-terminal fragment).
50 μL
Primary
Human, Mouse, Rat, Canine, Monkey, Avian, Zebrafish, Marsupial
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.