Learn More
Invitrogen™ FANCD2 Polyclonal Antibody, DyLight™ 650

Description
In Western blot, this antibody should recognize a band at ~166 kDa (post-translationally modified form). Additional bands may be seen at lower molecular weights. In immunofluorescence, this has been tested in human MMC and IR-treated MEF cells.
Specifications
Specifications
| Antigen | FANCD2 |
| Applications | ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | DyLight 650 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | FANCD2 |
| Gene Accession No. | Q6IV68, Q80V62, Q9BXW9 |
| Gene Alias | 2410150O07Rik; AU015151; BB137857; DKFZp762A223; FA4; FACD; FACD2; FAD; FAD2; FA-D2; FANCD; FANCD2; Fanconi anemia complementation group D2; Fanconi anemia D2 protein; Fanconi anemia group D2 protein; Fanconi anemia group D2 protein homolog; Fanconi anemia, complementation group D2; FLJ23826; Protein FACD2 |
| Gene Symbols | FANCD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.