Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2 Rabbit anti-Human, Alexa Fluor 488, Clone: 1290D, R&D Systems™

Catalog No. IC9369G025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC9369G025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC9369G025 Supplier R&D Systems Supplier No. IC9369G025
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FANCD2 Monoclonal specifically detects FANCD2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen FANCD2
Applications Flow Cytometry
Classification Monoclonal
Clone 1290D
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9BXW9
Gene Alias DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Host Species Rabbit
Immunogen E. coli-derived recombinant human FANCD2 Glu11-Glu230 Accession # Q9BXW9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2177
Test Specificity Detects human FANCD2 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.