Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fancd2os Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139940 Supplier Invitrogen™ Supplier No. PA5139940
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: WTPLDESFQW LRHTTPTPSS KHPFKASPCF PHTPSDLEVQ LCFQEVTLVL DSPFLESGVS PKLPCHTSEL RTMNNKGLVR K Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 88%.

FANCD2OS is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fancd2os
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FANCD2OS
Gene Accession No. Q96PS1
Gene Alias 4931417G12Rik; C20H3orf24; C22H3orf24; C3orf24; Fancd2 opposite strand; FANCD2 opposite strand protein; FANCD2OS; Fanconi anemia group D2 protein opposite strand transcript protein; HSD19; RGD1565997; uncharacterized protein C3orf24 homolog; Unknown (protein for MGC:133872)
Gene Symbols FANCD2OS
Host Species Rabbit
Immunogen Recombinant protein fragment of Human FANCD2OS (Product #RP-109538)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115795
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.