Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fancd2os Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569926 Supplier Invitrogen™ Supplier No. PA569926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

FANCD2OS is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fancd2os
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FANCD2OS
Gene Accession No. 0
Gene Alias 4931417G12Rik; C20H3orf24; C22H3orf24; C3orf24; Fancd2 opposite strand; FANCD2 opposite strand protein; FANCD2OS; Fanconi anemia group D2 protein opposite strand transcript protein; HSD19; RGD1565997; uncharacterized protein C3orf24 homolog; Unknown (protein for MGC:133872)
Gene Symbols FANCD2OS
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human Fancd2os.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500294
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.