Learn More
FANCE Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302126A

Description
The recommended shelf life for this product is 1 year from date of receipt.
As part of the Fanconi anemia (FA) complex functions in DNA cross-links repair. Required for the nuclear accumulation of FANCC and provides a critical bridge between the FA complex and FANCD2.Specifications
FANCE | |
Polyclonal | |
Unconjugated | |
FANCE | |
FAE; FACE; | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2178 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9HB96 | |
FANCE | |
Between 500 and 550 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.