Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FANCF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598907
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598907 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598907 Supplier Invitrogen™ Supplier No. PA598907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FANCF Polyclonal Antibody for Western Blot, ELISA

The Fanconi anemia complementation group currently includes FANCA, FANCB, FANCC, FANCD1 , FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ , FANCL, FANCM and FANCN . The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group F.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FANCF
Gene Accession No. Q9NPI8
Gene Alias A730016A17; FAF; FANCF; Fanconi anemia complementation group F; Fanconi anemia group F protein; Fanconi anemia, complementation group F; MGC126856; Protein FACF; RGD1561456
Gene Symbols FANCF
Host Species Rabbit
Immunogen Recombinant Human Fanconi anemia group F protein (295-341AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2188
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.