Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FANCG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111215
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111215 Supplier Invitrogen™ Supplier No. PA5111215
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSVLRASCLL PELLSALHRL VGLQAALWLS ADRLGDLALL LETLNGSQSG ASKDLLLLLK TWSPPAEELD APLTLQDAQG LKDVLLTAFA YRQGLQELIT GNPDKALSSL HEAASGLCPR PVLVQVYTAL GSCHRKMG Highest antigen sequence indentity to the following orthologs: Mouse - 72%, Rat - 71%.

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. FANCG is the protein for complementation group G.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCG
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FANCG
Gene Accession No. O15287
Gene Alias AU041407; DNA repair protein XRCC9; FAG; Fancg; Fanconi anemia complementation group G; Fanconi anemia group G protein; Fanconi anemia group G protein homolog; Fanconi anemia, complementation group G; Protein FACG; truncated Fanconi anemia group G protein; X-ray repair complementing defective repair in Chinese hamster cells 9; X-ray repair, complementing defective, in Chinese hamster, 9; Xrcc9
Gene Symbols FANCG
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human FANCG. Recombinant protein control fragment (Product #RP-106085).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.