Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCI Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300243FR
Description
FANCI Polyclonal antibody specifically detects FANCI in Human samples. It is validated for Western BlotSpecifications
FANCI | |
Polyclonal | |
Western Blot | |
Fanconi anemia group I protein, Fanconi anemia, complementation group I, KIAA1794FLJ10719, Protein FACI | |
A synthetic peptide mapping to a region between residues 200 and 250 of human KIAA1794/FLJ10719 using the numbering given in TrEMBL entry Q9NVI1 (GeneID 55215). | |
0.1 mL | |
Neuroscience, Stem Cell Markers | |
55215 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction