Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCI Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB300243
Description
FANCI Polyclonal specifically detects FANCI in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
FANCI | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Fanconi anemia group I protein, Fanconi anemia, complementation group I, KIAA1794FLJ10719, Protein FACI | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2500-1:10000, Immunoprecipitation 1- 4 ug/mg lysate | |
Q9NVI1 | |
FANCI | |
A synthetic peptide mapping to a region between residues 200 and 250 of human KIAA1794/FLJ10719 using the numbering given in TrEMBL entry Q9NVI1 (GeneID 55215). | |
100 μL | |
Neuroscience, Stem Cell Markers | |
55215 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction