Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ FANCI, (G4270), Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE1817 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1817 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1817 Supplier MilliporeSigma™ Supplier No. ABE1817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fanconi anemia group I protein (UniProt Q9NVI1; also known as Protein FACI) is encoded by the FANCI (also known as KIAA1794) gene (Gene ID 55215) in human. It plays a vital role in the repair of DNA double-strand breaks by homologous recombination and in the repair of interstrand DNA cross-links (ICLs) by promoting FANCD2 monoubiquitination by FANCL and participating in recruitment to DNA repair sites. It is also required for maintenance of chromosomal stability. FACI is reported to participate in S phase and G2 phase checkpoint activation upon DNA damage. It is monoubiquitinated by FANCL on Lys-523 during S phase and upon genotoxic stress and is deubiquitinated by USP1 as cells enter G2/M or once the DNA repair is completed. Mutations in the FANCI gene are known to cause Fanconi anemia, a developmental and cancer-predisposition syndrome. Ref: Smogorzewska A., et al. (2007) Cell 129, 289-301.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCI, (G4270)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene FANCI, KIAA1794
Gene Accession No. Q9NVI1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to an internal sequence within the N-terminal half of human FANCI.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001106849
Test Specificity This polyclonal antibody detected the expected target band only in lysate from FANCI-tranfected, but not untransfected FANCI-deficient EUFA1301 cells.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.