Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCI, p Ser556 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260780
Description
FANCI Polyclonal specifically detects FANCI in Human, Bovine, Goat, Primate, Sheep samples. It is validated for Western Blot.Specifications
FANCI (p Ser556) | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Fanconi anemia group I protein, Fanconi anemia, complementation group I, KIAA1794FLJ10719, Protein FACI | |
Rabbit | |
150 kDa | |
100 μL | |
Primary | |
Human, Bovine, Goat, Primate, Sheep | |
IgG |
Western Blot | |
Unconjugated | |
10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative | |
FANCI | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser556 conjugated to KLH. Accession # Q9NVI1 | |
Antigen Affinity-purified | |
Neuroscience, Stem Cell Markers | |
55215 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction