Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FANCI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116991 Supplier Invitrogen™ Supplier No. PA5116991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, 293T, MCF7 Immunogen sequence: DQDVEVEKTN HFAIVNLRTA APTVCLLVLS QAEKVLEEVD WLITKLKGQV SQETLSEEAS SQATLPNQPV EKAIIMQLGT LLTFFHELVQ TALPSGSCVD TLLKDLCKMY TTLTALVRYY LQVCQSSGGI PKNMEKLVKL SGSHLTPLCY SFISYVQNKS KSLNYTGEKK EKPAAVATAM ARVLRETKPI PNLIFAIEQY EKFLIHLSKK SKVNLMQHMK LSTSRDFKIK GNILDMVLRE DGEDENEEGT ASEHGGQNKE PAKKKRKK.

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group I. Alternative splicing results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCI
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FANCI
Gene Accession No. Q9NVI1
Gene Alias FANCI; Fanconi anemia complementation group I; Fanconi anemia group I protein; Fanconi anemia group I protein homolog; Fanconi anemia, complementation group I; KIAA1794; Protein FACI
Gene Symbols FANCI
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1061-1328 of human FANCI (NP_0011068491).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55215
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.