Learn More
Invitrogen™ FANCL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593235
Description
Immunogen sequence: MAVTEASLLR QCPLLLPQNR SKTVYEGFIS AQGRDFHLRI VLPEDLQLKN ARLLCSWQLR TILSGYHRIV QQRMQHSPDL MSFMMELKML LEVALKNRQE LYALPPPPQF YSSLIEEIGT LGWDKLVYAD TCFSTIKLKA EDASGREHLI TLKLKAKYPA ESPDYFVDFP VPFCASWTPQ SSLISIYSQF LAAIESLKAF WDVMDEIDEK TWVLEPEKPP RSATARRIAL GNNVSINIEV DPRHPTMLPE CFFLG; Positive Samples: Mouse thymus; Cellular Location: Cytoplasm, Nucleus.
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group L. Alternative splicing results in two transcript variants encoding different isoforms.
Specifications
FANCL | |
Polyclonal | |
Unconjugated | |
FANCL | |
2010322C19Rik; AW554273; B230118H11Rik; E3 ubiquitin-protein ligase FANCL; FA complementation group L; FAAP43; FANCL; Fanconi anemia complementation group L; fanconi anemia group L protein; Fanconi anemia group L protein homolog; Fanconi anemia, complementation group L; fanconi anemia-associated polypeptide of 43 kDa; FLJ10335; gcd; germ cell deficient; PHD finger protein 9; Phf9; Pog; proliferation of germ cells protein; RING-type E3 ubiquitin transferase FANCL | |
Rabbit | |
Affinity Chromatography | |
RUO | |
67030 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CR14 | |
FANCL | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-255 of human FANCL (NP_0605322). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.