Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576229 Supplier Thermo Scientific Supplier No. PA576229
Add to Cart
Add to Cart

FANCM Polyclonal Antibody

FANCM Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCM
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene FANCM
Gene Accession No. Q8IYD8
Gene Alias AI427100; ATP-dependent RNA helicase FANCM; C730036B14Rik; D12Ertd364e; FAAP250; FANCM; Fanconi anemia complementation group M; Fanconi anemia group M protein; Fanconi anemia group M protein homolog; Fanconi anemia, complementation group M; fanconi anemia-associated polypeptide of 250 kDa; Kiaa1596; Protein FACM; Protein Hef ortholog; RGD1307897
Gene Symbols FANCM
Host Species Rabbit
Immunogen Recombinant fusion protein of human FANCM.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57697
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.