Learn More
FANCM Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302637A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
ATPase required for FANCD2 ubiquitination, a key reaction in DNA repair. Binds to ssDNA but not to dsDNA. Recruited to forks stalled by DNA interstrand cross-links, and required for cellular resistance to such lesions.Specifications
FANCM | |
Polyclonal | |
Unconjugated | |
FANCM | |
Fanconi anemia group M protein, Protein FACM, ATP-dependent RNA helicase FANCM, Fanconi anemia-associated polypeptide of 250 kDa, FAAP250, Protein Hef ortholog, FANCM, KIAA1596 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57697 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8IYD8 | |
FANCM | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.