Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287433
Description
FANK1 Polyclonal specifically detects FANK1 in Human samples. It is validated for Western Blot.Specifications
FANK1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
1700007B22Rik, fibronectin type 3 and ankyrin repeat domains 1, fibronectin type 3 and ankyrin repeat domains protein 1, fibronectin type III and ankyrin repeat domains 1 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human FANK1 | |
100 μL | |
Developmental Biology | |
92565 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction