Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAP Mouse anti-Mouse, Clone: 73.3, MilliporeSigma™

Catalog No. MABC1145MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1145MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1145MI Supplier MilliporeSigma Supplier No. MABC1145
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect fibroblast activation protein using this mouse monoclonal Anti-FAP Antibody, clone 73.3. It has been tested in Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAP
Applications Flow Cytometry
Classification Monoclonal
Clone 73.3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Fap
Host Species Mouse
Immunogen FAP-expressing 3T3 cells.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_032012
Test Specificity Clone 73.3 specifically detects fibroblast activation protein (FAP) in 3T3 cells overexpressing FAP.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.