Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599458
Description
Antibody detects endogenous levels of total FAP1.
The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family. It is selectively expressed in reactive stromal fibroblasts of epithelial cancers, granulation tissue of healing wounds, and malignant cells of bone and soft tissue sarcomas. This protein is thought to be involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development, tissue repair, and epithelial carcinogenesis.
Specifications
FAP | |
Polyclonal | |
Unconjugated | |
FAP | |
170 kDa melanoma membrane-bound gelatinase; Antiplasmin-cleaving enzyme FAP, soluble form; APCE; Dipeptidyl peptidase FAP; DPPIV; Fap; FAPA; FAPalpha; fibroblast activation protein; fibroblast activation protein alpha; fibroblast activation protein, alpha; gelatine degradation protease FAP; Integral membrane serine protease; post-proline cleaving enzyme; Prolyl endopeptidase FAP; seprase; Serine integral membrane protease; SIMP; Surface-expressed protease | |
Rabbit | |
Affinity chromatography | |
RUO | |
14089, 192203, 2191 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P97321, Q12884 | |
FAP | |
A synthesized peptide derived from human FAP(Accession Q12884), corresponding to amino acid residues W346-E396. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction