Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAPAS Series 5 OC Pesticide Mix 1, Restek™
SDP

Catalog No. 06720010 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-720-010 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-720-010 Supplier Restek Corporation Supplier No. 32412
  • Equal concentration of all compounds
  • Suitable for GC-MS analysis
Compounds:

  • Aldrin (309-00-2)
  • α-BHC (319-84-6)
  • β-BHC (319-85-7)
  • γ-BHC (Lindane) (58-89-9)
  • cis-Chlordane (5103-71-9)
  • trans-Chlordane (5103-74-2)
  • 4,4'-DDD (72-54-8)
  • 4,4'-DDE (72-55-9)
  • 2,4'-DDT (789-02-6)
  • 4,4'-DDT (50-29-3)
  • Dieldrin (60-57-1)
  • α-Endosulfan (I) (959-98-8)
  • β-Endosulfan (II) (33213-65-9)
  • Endosulfan sulfate (1031-07-8)
  • Endrin (72-20-8)
  • Heptachlor (76-44-8)
  • Heptachlor epoxide (isomer B) (1024-57-3)
  • Hexachlorobenzene (118-74-1)
  • Oxychlordane (27304-13-8)

Specifications

Chemical Name or Material Acetone
Packaging Ampul
CAS Max % 99.810000
Ignition Point 465°C
Odor Strong
Vapor Density 2.0 (Air = 1)
Specific Gravity 0.7845g/cm3 at 25°C
Molecular Formula C3H6O
Concentration 100 μg/mL each in acetone
MDL Number MFCD00008765
Melting Point -95.4°C
Solubility Completely soluble
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Quantity 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.