Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAPP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518211
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518211 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518211 Supplier Invitrogen™ Supplier No. PA518211
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000. Preliminary analysis showed no signal in Western Blot.

Cargo transport protein that is required for apical transport from the Golgi complex. Transports AQP2 from the trans-Golgi network (TGN) to sites of AQP2 phosphorylation. Mediates the non-vesicular transport of glucosylceramide (GlcCer) from the trans-Golgi network (TGN) to the plasma membrane and plays a pivotal role in the synthesis of complex glycosphingolipids. Binding of both phosphatidylinositol 4-phosphate (PIP) and ARF1 are essential for the GlcCer transfer ability. Also required for primary cilium formation, possibly by being involved in the transport of raft lipids to the apical membrane, and for membrane tubulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAPP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PLEKHA8
Gene Accession No. Q96JA3
Gene Alias 6330400G01; AA517676; BC052360; FAPP2; Fapp-2; hFAPP2; MGC3358; PH domain-containing family A member 8; phosphatidylinositol-four-phosphate adapter protein 2; phosphoinositol 4-phosphate adapter protein 2; pleckstrin homology domain containing A8; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8; pleckstrin homology domain-containing family A member 8; PLEKHA8; RGD1563940; Serologically defined breast cancer antigen NY-BR-86
Gene Symbols PLEKHA8
Host Species Goat
Immunogen Synthetic peptide sequence (DIQTALRNPTENT) corresponding to the C-terminus amino acids of PLEKHA8 (aa 428-434).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84725
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.