Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAR1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9437R

Description
MLSTD2 catalyzes the reduction of saturated fatty acyl-CoA with chain length C16 or C18 to fatty alcohols.Specifications
FAR1 | |
Polyclonal | |
Unconjugated | |
FAR1 | |
2600011M19Rik; 2900034E22Rik; 3732409C05Rik; AI850429; BOS_14472; FAR1; fatty acyl CoA reductase 1; Fatty acyl-CoA reductase 1; LOC100351559; male sterility domain containing 2; Male sterility domain-containing protein 2; MLSTD2; PFCRD; SDR10E1; short chain dehydrogenase/reductase family 10E, member 1; UNQ2423/PRO4981 | |
Rabbit | |
Protein A | |
RUO | |
84188 | |
-20°C | |
Liquid |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8WVX9 | |
FAR1 | |
KLH conjugated synthetic peptide derived from human FAR1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction