Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Farnesol, 96%, mixture of isomers

Catalog No. p-133122
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle

CAS: 4602-84-0 | C15H26O | 222.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4602-84-0
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00002918,MFCD00002918,MFCD00002918,MFCD00002918
InChI Key CRDAMVZIKSXKFV-YFVJMOTDSA-N
Synonym e,z-farnesol, 2-trans,6-cis-farnesol, 2e,6z-farnesol, trans,cis-farnesol, farnesol, 2e,6z-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, farnesyl alcohol, e,z-3,7,11-trimethyl-2,6,10-dodecatrien-1-ol, 2-trans,6-cis-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
PubChem CID 1549109
ChEBI CHEBI:35966
IUPAC Name (2E,6Z)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CO

Specifications

Color Colorless to Yellow
Density 0.8865g/mL
Boiling Point 149°C (4.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (sum of geometric isomers) (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.492
Quantity 25 mL
Beilstein 01, III, 2040
Specific Gravity 0.8865
Merck Index 15, 3975
Solubility Information Solubility in water: immiscible
Formula Weight 222.37
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Farnesol, 96%
Show More Show Less
EINECSNumber 225-004-1
RTECSNumber JR4979000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.