Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Farnesylacetone, mixture of isomers, 97%

Catalog No. p-7024485
Click to view available options
Quantity:
50 g
250 g

CAS: 762-29-8 | C18H30O | 262.437 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-29-8
Molecular Formula C18H30O
Molecular Weight (g/mol) 262.437
MDL Number MFCD00036517
InChI Key LTUMRKDLVGQMJU-IUBLYSDUSA-N
Synonym farnesyl acetone, 5e,9e-6,10,14-trimethylpentadeca-5,9,13-trien-2-one, farnesylacetone, e,e-farnesylacetone, unii-3s0g4n267h, e,e-6,10,14-trimethylpentadeca-5,9,13-trien-2-one, trans,trans-farnesylacetone, 5,9,13-pentadecatrien-2-one, 6,10,14-trimethyl-, e,e, 2,6,10-trimethyl-2,6,10-pentadecatrien-14-one, 5,9,13-pentadecatriene-2-one, 6,10,14-trimethyl
PubChem CID 1711945
ChEBI CHEBI:67252
IUPAC Name (5E,9E)-6,10,14-trimethylpentadeca-5,9,13-trien-2-one
SMILES CC(=CCCC(=CCCC(=CCCC(=O)C)C)C)C

Specifications

Density 0.88
Flash Point >110°C (230°F)
Odor Pleasant
Refractive Index 1.481
Quantity 50 g
Beilstein 1781239
Formula Weight 262.44
Percent Purity 97%
Chemical Name or Material Farnesylacetone, mixture of isomers
EINECSNumber 212-097-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.