Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FARP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP276803 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276803 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276803 Supplier Novus Biologicals Supplier No. NBP276803
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

FARP1 Polyclonal specifically detects FARP1 in Human samples. It is validated for Flow Cytometry, Peptide ELISA, Immunofluorescence.

Specifications

Antigen FARP1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 10 μ/mL, Peptide ELISA Detection Limit 1:32000, Immunofluorescence 10 μ/mL
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_005757, NP_001273768.1, NP_001001715.2
Gene Alias CDEPPleckstrin homology domain-containing family C member 2, Chondrocyte-derived ezrin-like protein, FERM, RhoGEF (ARHGEF) and pleckstrin domain protein 1 (chondrocyte-derived), FERM, RhoGEF and pleckstrin domain-containing protein 1, MGC87400, PLEKHC2PH domain-containing family C member 2
Gene Symbols FARP1
Host Species Goat
Immunogen Peptide with sequence GEIEQRPTPGSRL-C, from the N Terminus of the protein sequence according to NP_005757.1; NP_001001715.2; NP_001273768.1.
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 10160
Test Specificity This antibody is expected to recognize isoform 1 (NP_005757.1), isoform 2 (NP_001001715.2), and isoform 3 (NP_001273768.1)
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.