Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FARP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548937
Description
Predicted to react with chicken based on sequence homology.
Functions as guanine nucleotide exchange factor for RAC1. May play a role in semaphorin signaling. Plays a role in the assembly and disassembly of dendritic filopodia, the formation of dendritic spines, regulation of dendrite length and ultimately the formation of synapses.
Specifications
FARP1 | |
Polyclonal | |
Unconjugated | |
FARP1 | |
CDEP; chondrocyte-derived ezrin-like protein; FARP1; FARP1-IT1; FERM, ARH/RhoGEF and pleckstrin domain protein 1; FERM, ARHGEF and pleckstrin domain-containing protein 1; FERM, RhoGEF (ARHGEF) and pleckstrin domain protein 1 (chondrocyte-derived); FERM, RhoGEF and pleckstrin domain-containing protein 1; PH domain-containing family C member 2; pleckstrin homology domain-containing family C member 2; PLEKHC2; PPP1R75; protein phosphatase 1, regulatory subunit 75 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10160 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9Y4F1 | |
FARP1 | |
KLH conjugated synthetic peptide between 62-90 amino acids from the N-terminal region of human FARP1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction