Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590317 Supplier Invitrogen™ Supplier No. PA590317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVGSALRRGA HAYVYLVSKA SHISRGHQHQ AWGSRPPAAE CATQRAPGSV VELLGKSYPQ DDHSNLTRKV LTRVGRNLHN QQHHPLWLIK ERVKEHFYKQ YVGRFGTPLF SVYDNLSPVV TTWQNFDSLL IPADHPSRKK GDNYYLNRTH MLRAHTSAHQ WDLLHAGLDA FLVVGDVYRR DQIDSQHYPI FHQLEAVRLF SKHELFAGIK DGESLQLFEQ SSRSAHKQET HTMEAVKLVE FDLKQTLTRL; Positive Samples: A549, rat kidney; Cellular Location: Mitochondrion matrix.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS2 is a phenylalanine-tRNA synthetase (PheRS) localized to the mitochondrion which consists of a single polypeptide chain, unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and in the eukaryotic cytoplasm. Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS1 encodes a mitochondrially-located phenylalanine-tRNA synthetase (PheRS) which consists of a single polypeptide chain unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and the eukaryotic cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FARS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FARS2
Gene Accession No. O95363, Q6AYQ3
Gene Alias COXPD14; dJ236A3.1 (phenylalanine-tRNA synthetase); dJ520B18.2; dJ520B18.2 (FARS1 (phenylalanine-tRNA synthetase)); Fars1; FARS2; HSPC320; mitochondrial PHERS; pheny; phenylalanine translase; phenylalanine tRNA ligase 2, mitochondrial; phenylalanine--tRNA ligase, mitochondrial; phenylalanine-tRNA synthetase 1 (mitochondrial); phenylalanine-tRNA synthetase 2 (mitochondrial); Phenylalanyl-tRNA synthetase; phenylalanyl-tRNA synthetase 2, mitochondrial; phenylalanyl-tRNA synthetase, mitochondrial; phenylalanyl-tRNA synthetase, mitochondrial; phenylalanine--tRNA ligase, mitochondrial; pheRS
Gene Symbols FARS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human FARS2 (NP_0065581).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10667, 306879
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.